4'-Aminoacetophenone ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A0251-25G A0251-100G
TCA0251-025GEA 102.7 CAD
TCA0251-025G TCA0251-500G TCA0251-100G
4'-Aminoacetophenone ≥98.0% (by titrimetric analysis)
4'-Aminoacetophenone

CAS Number: 99-92-3
MDL Number: MFCD00007896
Molecular Formula: C8H9NO
Molecular Weight: 135.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 293
Melting point (°C): 107

Formula: C₈H₉NO
Boiling Pt: 293 °C
Melting Pt: 107 °C
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007896
CAS Number: 99-92-3
EINECS: 202-801-2
Merck Index: 13,00412
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8bfb2b7de4097862557cc720cd5dbe26cecde2c&vid=f8bfb2b7de4097862557cc720cd5dbe26cecde2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR