Butyl acetate ≥99.0%

Supplier: TCI America
Danger

Synonyms: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate

A0228-500ML
TCA0228-500MLEA 99.68 CAD
TCA0228-500ML
Butyl acetate ≥99.0%
Butyl acetate

[for Spectrophotometry]
CAS Number: 123-86-4
MDL Number: MFCD00009445
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Melting point (°C): -78
Flash Point (°C): 28
Specific Gravity (20/20): 0.88


Product Categories: Analytical Science, Solvents for Spectrophotometry
Other Product Names / Synonyms: Acetic Acid Butyl Ester

Formula: CH₃COO(CH₂)₃CH₃
MW: 116.16 g/mol
Boiling Pt: 126
Melting Pt: -76
Density: 0.88 (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00009445
CAS Number: 123-86-4
EINECS: 204-658-1
UN: 1123
ADR: 3 III
Merck Index: 13,01534
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1326c53753bc09b081f43a9e3ef32314f05439b1&vid=d87afc796696db6495557b13c89082603d08ca32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR