Ethyl allophanate ≥98.0% (by total nitrogen basis)

Supplier: TCI America

A0213-25G
TCA0213-025GEA 680.98 CAD
TCA0213-025G
Ethyl allophanate ≥98.0% (by total nitrogen basis)
Ethyl allophanate

CAS Number: 626-36-8
MDL Number: MFCD00047870
Molecular Formula: C4H8N2O3
Molecular Weight: 132.12
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 190


Other Product Names / Synonyms: Allophanic Acid Ethyl Ester

Formula: C₄H₈N₂O₃
Melting Pt: 190 °C
MDL Number: MFCD00047870
CAS Number: 626-36-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63c11f4f87cfd64adfcaf8ab16d342ab8a9d1b57&vid=63c11f4f87cfd64adfcaf8ab16d342ab8a9d1b57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR