1-Acetyl-3-methylurea ≥98.0%

Supplier: TCI America

A0101-25G
TCA0101-025GEA 221.2 CAD
TCA0101-025G
1-Acetyl-3-methylurea ≥98.0%
1-Acetyl-3-methylurea

CAS Number: 623-59-6
MDL Number: MFCD00026167
Molecular Formula: C4H8N2O2
Molecular Weight: 116.12
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 180

Formula: C₄H₈N₂O₂
MW: 116.12 g/mol
Melting Pt: 180 °C
MDL Number: MFCD00026167
CAS Number: 623-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7c56bdfbd69ed9ff3eeb9adcb85adb96b24c901&vid=b7c56bdfbd69ed9ff3eeb9adcb85adb96b24c901&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR