N-Acetyl-DL-methionine ≥98.0%

Supplier: TCI America

Synonyms: Ac-DL-Met-OH

A0100-25G A0100-500G
TCA0100-025GEA 44.24 CAD
TCA0100-025G TCA0100-500G
N-Acetyl-DL-methionine ≥98.0%
N-Acetyl-DL-methionine

CAS Number: 1115-47-5
MDL Number: MFCD00008681
Molecular Formula: C7H13NO3S
Molecular Weight: 191.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 116


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-DL-Met-OH

Formula: C₇H₁₃NO₃S
Melting Pt: 116 °C
MDL Number: MFCD00008681
CAS Number: 1115-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7edb33a53efd928a09b409a4d0fbc32212d59fa2&vid=7edb33a53efd928a09b409a4d0fbc32212d59fa2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR