Acetone oxime ≥98.0%

Supplier: TCI America

A0065-25G A0065-500G
TCA0065-025GEA 36.34 CAD
TCA0065-025G TCA0065-500G
Acetone oxime ≥98.0%
Acetone oxime

CAS Number: 127-06-0
MDL Number: MFCD00002118
Molecular Formula: C3H7NO
Molecular Weight: 73.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 133
Melting point (°C): 62

Formula: C₃H₇NO
Boiling Pt: 133 °C
Melting Pt: 62 °C
Density: 1.045 g/cm³ (20 °C)
MDL Number: MFCD00002118
CAS Number: 127-06-0
EINECS: 204-820-1
UN: 1325
ADR: 4.1,III
Merck Index: 13,00077
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd4d42bdf1e7de2599d280905be572fa1fe5a744&vid=bd4d42bdf1e7de2599d280905be572fa1fe5a744&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR