2'-Methylacetanilide ≥98.0%

Supplier: TCI America

Synonyms: o-Acetotoluidide, o-Acetotoluidid, N-(o-Tolyl)acetamide, N-(2-Methylphenyl)acetamide

A0063-25G
TCA0063-025GEA 62.88 CAD
TCA0063-025G
2'-Methylacetanilide ≥98.0%
2'-Methylacetanilide

CAS Number: 120-66-1
MDL Number: MFCD00014961
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 296
Melting point (°C): 110


Other Product Names / Synonyms: N-Acetyl-o-toluidine, 2'-Methylacetanilide

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Boiling Pt: 296 °C
Melting Pt: 110
Density: 1.168 g/cm³ (15 °C)
MDL Number: MFCD00014961
CAS Number: 120-66-1
EINECS: 204-414-4
Merck Index: 12,00074
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e341f742325076fb8fcd2dca12fdca146177fcdf&vid=e341f742325076fb8fcd2dca12fdca146177fcdf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR