Acetophenone ≥98.5%

Supplier: TCI America
Warning

A0061-25G A0061-500G
TCA0061-025GEA 37.62 CAD
TCA0061-025G TCA0061-500G
Acetophenone ≥98.5%
Acetophenone

CAS Number: 98-86-2
MDL Number: MFCD00008724
Molecular Formula: C8H8O
Molecular Weight: 120.15
Purity/Analysis Method: >98.5% (GC)
Form: Clear Liquid
Boiling point (°C): 202
Flash Point (°C): 80
Freezing point (°C): 19
Specific Gravity (20/20): 1.03


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: Acetylbenzene, Methyl Phenyl Ketone

Formula: C₈H₈O
Boiling Pt: 202 °C
Melting Pt: 20 °C
Density: 1.03 (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00008724
CAS Number: 98-86-2
EINECS: 202-708-7
UN: 3334
ADR: 9,
Merck Index: 13,00074
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ae1cd68496c86406a95b5ffc18f35bd3c6ed153&vid=e2673dd23819859931f97bfc83cfe9fe0f5b38b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR