Claritas PPT® Grade Minerals Testing Kit, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
SPXMT-KIT
SXSPXMT-KITEA 523.86 CAD
SXSPXMT-KIT
Claritas PPT® Grade Minerals Testing Kit, SPEX CertiPrep
Standards Cationic Standards Multi-Element Standards
Claritas PPT® Grade Minerals Testing Kit for ICP-MS are designed for routinely analyzed minerals.
Kits come with six, 30 mL standards which include a nitric acid blank for easy dilution. Conveniently packaged in a sturdy, heavy-duty carton, these kits are perfect to store on a lab bench or in a cabinet.
The 30 mL standards ship non-hazardous, saving money on shipping costs. The smaller volume also allows for less hazardous waste, should the standard expire before its contents are used.