Phenanthrene-d10 Single-Component Organic Standard, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
S-3020
SXS-3020EA 114.46 CAD
SXS-3020
Phenanthrene-d10 Single-Component Organic Standard, SPEX CertiPrep
Standards Organic Standards
Single-Component organic standards.
Single-component standards are available as stock items. They are supplied in methanol, or in another solvent, where appropriate. Additional solvents include methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.
Concentrations other than these, as well as custom matrices, are readily available, but a minimum order of five ampules may be required in some cases.