Anti-GDF15 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
600-403-B10
RL600-403-B10EA 760.15 CAD
RL600-403-B10
Anti-GDF15 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol GDF15
Environmentally Preferable
Antigen name Growth Differentiation Factor 15
Antigen synonyms MIC-1|GDF-15|NAG-1|PTGFB|PDF|PLAB|MIC1
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human
Host Rabbit
Gene ID 9518
Isotype IgG
Western blot Yes
ELISA Yes
Epitope N-Terminal
Immunogen This affinity purified antibody was prepared by repeated immunizations with a synthetic peptide corresponding to a region near the amino terminal end of human NAG-1 Protein.  A residue of cysteine was added to facilitate coupling to KLH.
Format Antibody fraction
Form Lyophilized
Purification affinity-purified
Cross adsorption No
Storage buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
Storage temperature –20 °C
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR