FAME #12 Mix (5 components), Restek
Supplier: RESTEK CORP
35021
RK35021EA 85.33 CAD
RK35021
FAME #12 Mix (5 components), Restek
Standards Organic Standards
Contains: Chain, Compound (CAS#), % by Weight; C13:00 Methyl tridecanoate (1731-88-0), 20%; C15:00 Methyl pentadecanoate (7132-64-1), 20%; C17:00 Methyl heptadecanoate (1731-92-6), 20%; C19:00 Methyl nonadecanoate (1731-94-8), 20%; C21:00 Methyl heneicosanoate (6064-90-0), 20%