EPA method 505 Organohalide Pesticide Mixture, Restek
Supplier: RESTEK CORP
32024
RK32024EA 112.78 CAD
RK32024
EPA method 505 Organohalide Pesticide Mixture, Restek
Standards Environmental Analysis Standards
Mixture is packaged in methanol at a concentration of 200 µg/mL in a 1 mL ampoule.
  • Mixture contains the compounds Alachlor, Aldrin, Atrazine, [gamma]-BHC (Lindane), [alpha]-Chlordane, [gamma]-Chlordane, Dieldrin, Endrin, Heptachlor, Heptachlor Epoxide B, Hexachlorobenzene, Hexachlorocyclopentadiene, Methoxychlor, cis-Nonachlor, trans-Nonachlor, and Simazine
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).