CLP–Semi-Volatiles Base/Neutrals Surrogates (3/90 SOW), Restek
Supplier: RESTEK CORP
31072 31082 31002
RK31072EA 97.96 CAD
RK31072 RK31082 RK31002
CLP–Semi-Volatiles Base/Neutrals Surrogates (3/90 SOW), Restek
Standards Environmental Analysis Standards
Standards are packaged in methylene chloride at the stated concentration in an ampoule.
  • Mixture contains 1,2-Dichlorobenzene-d4, 2-Fluorobiphenyl, Nitrobenzene-d5, and p-Terphenyl-d14
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).