PA DEP UST Standard (11 components), Restek
Supplier: RESTEK CORP
30433
RK30433EA 71.89 CAD
RK30433
PA DEP UST Standard (11 components), Restek
Standards Petroleum Industry Standards
Contains: Benzene (71-43-2); 1,2-Dibromoethane (EDB) (106-93-4); 1,2-Dichloroethane (107-06-2); Ethylbenzene (100-41-4); Isopropyl benzene (cumene) (98-82-8); Methyl tert-butyl ether (MTBE) (1634-04-4); Naphthalene (91-20-3); Toluene (108-88-3); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3)