Chlorobenzene, Standard, 2000 μg/ml, Restek
Supplier: RESTEK CORP
30261
RK30261EA 53.72 CAD
RK30261
Chlorobenzene, Standard, 2000 μg/ml, Restek
Standards Organic Standards
Solvent packaged in purge-and-trap-grade methanol.
  • 2000 μg/mL in P&T methanol, 1 mL/ampoule
Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.