DW-VOC Mix #1 (8 components), Restek
Supplier: RESTEK CORP
30219
RK30219EA 109.26 CAD
RK30219
DW-VOC Mix #1 (8 components), Restek
Standards Environmental Analysis Standards
Contains: Benzene (71-43-2); Carbon tetrachloride (56-23-5); 1,4-Dichlorobenzene (106-46-7); 1,2-Dichloroethane (107-06-2); 1,1-Dichloroethene (75-35-4); 1,1,1-Trichloroethane (71-55-6); Trichloroethene (79-01-6); Vinyl chloride (75-01-4)