VOA Surrogate Spike Mix (3 components), Restek
Supplier: RESTEK CORP
30004
RK30004EA 73.9 CAD
RK30004
VOA Surrogate Spike Mix (3 components), Restek
Standards Organic Standards
Mix contains: 1-Bromo-4-fluorobenzene (BFB) (460-00-4), 1,2-Dichloroethane-d4 (17060-07-0) and Toluene-d8 (2037-26-5).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.