Double Taper Inlet Liner for Shimadzu 14A GCs Equipped with Split/Splitless Inlets, Restek
Supplier: Restek
20864
RK20864PK 282.35 CAD
RK20864
Double Taper Inlet Liner for Shimadzu 14A GCs Equipped with Split/Splitless Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.