Methane Cylinder Kit, Restek
Supplier: Restek
20197
RK20197EA 1303.95 CAD
RK20197
Methane Cylinder Kit, Restek
Gas Detection Systems
Setting the column flow rate by injecting methane and optimizing linear velocity is a preferred method for establishing reproducible retention times (ASTM Method E1510-93).
For optimizing linear velocity. The kit includes a Scotty® 14 cylinder containing 1% methane in helium, a MINICYL regulator, a syringe adapter, and a package of twenty-five septa for the adapter.
Accessories information: Replacement parts allow for continued maintenance of a methane cylinder kit, which is used for optimizing linear velocity.