Rtx®-5 GC Columns, Restek
Supplier: Restek
10227 10228 10240 10205 10209 10210 10211 10220 10222 10224 10226 10236 10238 40201 40203 10206 10208 10225 10229 10235 10237 10239 10241 10242 10221 10223-600 10243 10207 40210 40211 41201 41202 41204 10224-125 10220-127 10223-127 10224-128 10220-600 10220-124 10221-125 10222-126 10223-124 10225-126 10227-125 10238-124 10239-125 10222-129 10226-127 10240-129 40202 40212 10235-600
RK10227EA 2743.81 CAD
RK10227 RK10228 RK10240 RK10205 RK10209 RK10210 RK10211 RK10220 RK10222 RK10224 RK10226 RK10236 RK10238 RK40201 RK40203 RK10206 RK10208 RK10225 RK10229 RK10235 RK10237 RK10239 RK10241 RK10242 RK10221 RK10223-600 RK10243 RK10244 RK10207 10058-082 10058-084 10058-098 10058-100 10059-534 10059-916 10059-918 10059-920 10059-922 10059-924 10059-926 10803-458 10848-746 10849-184 10849-196 10849-198 10849-200 10849-214 10849-224 10849-666 10849-860 10850-296 10850-298 10850-300 10850-302 10850-304 10850-306 10850-312 10850-314 10850-576 10850-596 10850-600 10850-618 10850-620 10850-622 10850-792 10850-796 10850-826 10850-828 10850-842 10852-084 75849-944
Rtx®-5 GC Columns, Restek
Chromatography Columns
A low-polarity general-purpose column for the analysis of pesticides, PCBs, aromatic hydrocarbons, drugs, and essential oils.
  • Extremely low bleed
  • Low-polarity column
  • Internal dimensions and film thickness vary between models
Equivalent to USP G27, G36 phases. All residual catalysts and low molecular weight fragments are removed from the Rtx®-5 polymer, providing a tight mono-modal distribution and extremely low bleed. Phase composition: Crossbond® 5% diphenyl, 95% dimethylpolysiloxane.
Order Now
SPECIFICATIONS
Environmentally Preferable
Temperature limit –60...350 °C