Silver nitrate 0.01 N isopropanol

Supplier: Ricca Chemical

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

RSOD9000-1C
RCSOD90-32EA 1355.64 CAD
RCSOD90-32
Silver nitrate 0.01 N isopropanol
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1 II
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR