VeriSpec™ Chlorate - ClO₃₋ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010909-100N RV010909-250N RV010909-500N
RCRV010909100NEA 205.4 CAD
RCRV010909100N RCRV010909250N RCRV010909500N
VeriSpec™ Chlorate - ClO₃₋ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Standards Anionic Standards Chlorate Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility