VeriSpec® ICP Multi-Element QC Standard 7A, RICCA Chemical Company
Supplier: Ricca Chemical
RV010774-100N
RCRV010774100NEA 665.18 CAD
RCRV010774100N
VeriSpec® ICP Multi-Element QC Standard 7A, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
Seven component standard.
100 ppm: Al, B, Ba, NA
1000 ppm: K
500 ppm: Si
50 ppm: Ag
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility