VeriSpec® Mixed Anion Standard, 7 Component, Ricca Chemical Company
Supplier: Ricca Chemical
RV010740-100N
RCRV010740100NEA 848.46 CAD
RCRV010740100N
VeriSpec® Mixed Anion Standard, 7 Component, Ricca Chemical Company
Standards Anionic Standards Multi-Element Anioinic Standards
Certified reference material (CRM) for ion chromatography (IC)
0.5 ppm F-, PO₄³⁺; 1 ppm NO₂-, NO₃-, Br-; 12.5 ppm SO₄²⁺, Cl-
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility