VeriSpec® Mixed Cation Standard, 9 Components, Ricca Chemical Company
Supplier: Ricca Chemical
RV010730-100N
RCRV010730100NEA 889.54 CAD
RCRV010730100N
VeriSpec® Mixed Cation Standard, 9 Components, Ricca Chemical Company
Standards Cationic Standards Multi-Element Standards
Certified reference material (CRM) for ion chromatography (IC)
100 ppm Ba2+, Ca2+, K+, Li+, Mg2+, Mn2+, Na+, NH4+, Sr2+
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
SPECIFICATIONS
Environmentally Preferable