VeriSpec® ICP Multi-Element Standard 30, RICCA Chemical Company
Supplier: Ricca Chemical
RV010713-100N
RCRV010713100NEA 1867.56 CAD
RCRV010713100N
VeriSpec® ICP Multi-Element Standard 30, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
30 component standard.
1000 ppm: Ca
100 ppm: As, B, Be, Fe, Se, Zn
10 ppm: Ag, Al, Ba, Bi, Cd, Co, Cr, Cu, Ga, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Rb, Sr, Te, Tl, U, V
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
SPECIFICATIONS
Environmentally Preferable