VeriSpec® Strontium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010624-100N
RCRV010624-50NEA 141.32 CAD
RCRV010624-50N RCRV010624100N
VeriSpec® Strontium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Strontium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility