VeriSpec™ Erbium Standard for ICP/MS 10 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010586-100N
RCRV010586-50NEA 127.7 CAD
RCRV010586-50N RCRV010586100N
VeriSpec™ Erbium Standard for ICP/MS 10 ppm in 2% HNO₃
Standards Cationic Standards Erbium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility