VeriSpec® Cobalt Standard for ICP/MS 10 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010581-100N
RCRV010581100NEA 205.4 CAD
RCRV010581100N
VeriSpec® Cobalt Standard for ICP/MS 10 ppm in 2% HNO₃
Standards Cationic Standards Cobalt Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility