VeriSpec™ Ytterbium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010567-100N
RCRV010567100NEA 251.22 CAD
RCRV010567100N
VeriSpec™ Ytterbium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Ytterbium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility