VeriSpec® Samarium Standard for ICP 10 g/L in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010416-100N
RCRV010416100NEA 409.22 CAD
RCRV010416100N RCRV010416250N RCRV010416500N
VeriSpec® Samarium Standard for ICP 10 g/L in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Samarium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 10 g/L= 10,000 µg/mL