VeriSpec® Molybdenum Standard for ICP 1000 ppm in 4% NH₃, Ricca Chemical Company
Supplier: Ricca Chemical
RV010344-100N RV010344-250N RV010344-500N
RCRV010344-50NEA 115.58 CAD
RCRV010344-50N RCRV010344100N RCRV010344250N RCRV010344500N
VeriSpec® Molybdenum Standard for ICP 1000 ppm in 4% NH₃, Ricca Chemical Company
Standards Cationic Standards Molybdenum Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Order Now
SPECIFICATIONS
Environmentally Preferable