Tungsten Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010327-100N RV010327-250N RV010327-500N RV010428-100N RV010497-100N
RCRV010327100NEA 205.4 CAD
RCRV010327100N RCRV010327250N RCRV010327500N RCRV010428100N RCRV010497100N
Tungsten Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Tungsten Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml
Order Now
SPECIFICATIONS
Environmentally Preferable