VeriSpec® Cadmium (Cd) Standard for AAS 2 ppb in 1% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010123-50N
RCRV010123-50NEA 377.62 CAD
RCRV010123-50N
VeriSpec® Cadmium (Cd) Standard for AAS 2 ppb in 1% HNO₃, RICCA Chemical Company
Standards Cationic Standards Cadmium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility