Sodium carbonate 1.00 N

Supplier: Ricca Chemical
Danger

R7190000-4A R7190000-1A R7190000-500A
RC71901EA 249.64 CAD
RC71901 RC719032 RCR7190000500A
Sodium carbonate 1.00 N
Sodium carbonate

Clear, colorless liquid. Less than 1% sodium carbonate. Irritant.

Formula: Na₂CO₃
Melting Pt: ∼ 0°C
Density: 1.1
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Methyl Orange) 0.999-1.001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR