Aluminum ICP-MS Standard, 1 ml = 1 mg Al (1000 ppm Al)Al(NO₃)₃ in 3% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RMSAL1KN-100N
RCMSAL1KN-100EA 353.92 CAD
RCMSAL1KN-100
Aluminum ICP-MS Standard, 1 ml = 1 mg Al (1000 ppm Al)Al(NO₃)₃ in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Aluminum Standards