Xylenol orange indicator 0.1 % (w/v) in aqueous solution

Supplier: Ricca Chemical

R9386000-500A R9386000-120A
RC9386-16EA 254.38 CAD
RC9386-16 RC9386-4
Xylenol orange indicator 0.1 % (w/v) in aqueous solution
Xylenol orange
Formula: C₃₁H₂₈N₂Na₄O₁₃S
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7

Order Now

Specification Test Results

Appearance Clear, red, odorless
Suitability for Use Purple (Excess Metal) - Yellow (Excess EDTA)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR