L(+)-Potassium sodium tartrate 10% (w/v) in aqueous solution pH 5.2

Supplier: Ricca Chemical

R7540000-500A R7540000-1A
RC7540-16EA 183.28 CAD
RC7540-16 RC7540-32
L(+)-Potassium sodium tartrate 10% (w/v) in aqueous solution pH 5.2
L(+)-Potassium sodium tartrate
Formula: C₄H₄KNaO₆
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0500 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00065391
CAS Number: 304-59-6

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
pH at 25 °C (traceable to NIST SRM 185 & 186) 5.20 ± 0.05 pH at 25.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR