Sodium hydroxide 1.50 N

Supplier: Ricca Chemical

Synonyms: Caustic soda

R7451000-4A
RC7451-1EA 519.82 CAD
RC7451-1
Sodium hydroxide 1.50 N
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 1.500 ± 0.001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR