Sodium hydroxide 50% (w/w) in aqueous solution (760 g/L) analytical reagent

Supplier: Ricca Chemical
Danger

R7291000-4A R7291000-20F R7291000-500A R7291000-20E1 R7291000-30E
RC72911EA 486.64 CAD
RC72911 RC72915 RC729116 RC7291-5HP RCR7291000-30E
Sodium hydroxide 50% (w/w) in aqueous solution (760 g/L) analytical reagent
Sodium hydroxide
Formula: NaOH
Boiling Pt: ~100 °C
Melting Pt: 12 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay at 20 °C (traceable to NIST SRM 723) 50 - 52 % (w/w)
Chloride (Cl) 0.002% max
Heavy Metals (as Ag) 10 ppm (0.001%) max
Iron (Fe) 5 ppm (0.0005%) max
NH4OH Precipitate 0.01% max
Nickel (Ni) 5 ppm (0.0005%) max
Nitrogen Compounds (as N) 5 ppm (0.0005%) max
Phosphate (PO4) 5 ppm (0.0005%) max
Potassium (K) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR