Sodium chloride 0,9% (w/v) in aqueous solution isotonic saline, suitable for use as blood bank saline

Supplier: Ricca Chemical

Synonyms: Saline, NaCl, Sea salt, Halite

R7210000-4A R7210000-500A R7210000-10F R7210000-1A R7210000-20F
RC72101EA 131.14 CAD
RC72101 RC721016 RC721025 RC721032 RC72105
Sodium chloride 0,9% (w/v) in aqueous solution isotonic saline, suitable for use as blood bank saline
Sodium chloride

Isotonic saline, sub-micron filtered, low particle count, nonsterile.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 8.99 ± 0.01 g/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR