Silver nitrate 0.282 N

Supplier: Ricca Chemical

R7011000-4B R7011000-1B R7011000-20F
RC70111EA 3891.54 CAD
RC70111 RC701132 RC70115
Silver nitrate 0.282 N
Silver nitrate

1 mL = 10 mg Cl-

Formula: AgNO₃
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.2820 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR