Silver nitrate 0.0171 N

Supplier: Ricca Chemical

R6900000-4B R6900000-1B R6900000-20F
RC69001EA 338.12 CAD
RC69001 RC690032 RC69005
Silver nitrate 0.0171 N
Silver nitrate

1 mL = 1 mg NaCl

Formula: AgNO₃
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.01710 ± 0.00001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR