Diammonium hydrogen orthophosphate 30% (w/v) in aqueous solution (1 ml = 1 mg Phenol) APHA for magnesium

Supplier: Ricca Chemical
Warning

Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

R0689300-1A
RC689.30-32EA 300.2 CAD
RC689.30-32
Diammonium hydrogen orthophosphate 30% (w/v) in aqueous solution (1 ml = 1 mg Phenol) APHA for magnesium
di-Ammonium hydrogen orthophosphate
Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
Melting Pt: 185
Density: 1.2000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
Merck Index: 13,00546

Order Now

Specification Test Results

Appearance Clear, colorless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR