Silica Standard, 1 ml = 0.1 mg SiO₂, 100 ppm SiO₂ (46.7 ppm Si), Ricca Chemical Company
Supplier: Ricca Chemical
R6745000-500A
RC674516EA 126.4 CAD
RC674516
Silica Standard, 1 ml = 0.1 mg SiO₂, 100 ppm SiO₂ (46.7 ppm Si), Ricca Chemical Company
Standards Cationic Standards Silicium Standards