Ammonium molybdate solution Reagent 1 for phosphorus determination by the stannous chloride method (without extraction)

Supplier: Ricca Chemical
Danger

R0672000-500A
RC67216EA 216.46 CAD
RC67216
Ammonium molybdate solution Reagent 1 for phosphorus determination by the stannous chloride method (without extraction)
Ammonium molybdate solutions
Density: 1.3000 g/cm³
Storage Temperature: Ambient
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless to pale yellow-green liquid, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR