Potassium thiocyanate 0.100 N (N/10) in aqueous solution

Supplier: Ricca Chemical

R6580000-4A R6580000-1A
RC65801EA 320.74 CAD
RC65801 RC658032
Potassium thiocyanate 0.100 N (N/10) in aqueous solution
Potassium thiocyanate
Formula: KSCN
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.1000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR