Ammonium chloride 0.100 M (M/10) in aqueous solution analytical reagent

Supplier: Ricca Chemical

Synonyms: Ammonium muriate, Salmiac

R0627000-500A
RC62716EA 124.82 CAD
RC62716
Ammonium chloride 0.100 M (M/10) in aqueous solution analytical reagent
Ammonium chloride

For use as ammonia standard or as filling solution for sodium ion selective electrode.

Formula: NH₄Cl
MW: 53.49 g/mol
Melting Pt: 335
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.1000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR