Potassium hydroxide 1.00 N in isopropanol (IPA)

Supplier: Ricca Chemical

R6258000-4A
RC6258-1EA 502.44 CAD
RC6258-1
Potassium hydroxide 1.00 N in isopropanol (IPA)
Potassium hydroxide
Formula: KOH
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear to slightly hazy, colorless, Isopropanol odor
Assay at 20 °C (traceable to NIST SRM 723) 1.000 ± 0.005 N at 20.0 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR